2,4,6-Trimethylphenol ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A15041.14
A15041.22
A15041.14EA
2480
CZK
A15041.14
A15041.22
2,4,6-Trimethylphenol ≥98%
2,4,6-Trimethylphenol
Purity: 98%
CAS No.: 527-60-6
Molecular Formula: C9H12O
Formula Weight: 136.20
Melting Point: 70-74°
Boiling Point: 220-221°
UN No.: 2430
MDL No.: MFCD00002235
Vzorec:
C₉H₁₂O MW: 136,19 g/mol Bod varu: 220…221 °C Bod tání: 70…74 °C Hustota: 1,046 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002235 CAS číslo: 527-60-6 EINECS: 208-419-2 UN: 2430 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...