Tisk…

2-Bromomesitylene ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,4,6-Trimethylbromobenzene, 2-Bromo-1,3,5-trimethylbenzene, Mesityl bromide, 2-Bromomesitylene

A12277.18 A12277.22 A12277.30 A12277.36
A12277.18EA 1710 CZK
A12277.18 A12277.22 A12277.30 A12277.36
2-Bromomesitylene ≥99%
2-Bromomesitylene

Purity: 99% CAS No.: 576-83-0 Molecular Formula: C9H11Br Formula Weight: 199.10 Flash Point: 96°(204°F) Refractive Index: 1.5520 Melting Point: 2° Boiling Point: 224-226° Beilstein Reference: 1907245 MDL No.: MFCD00000073

Vzorec: C₉H₁₁Br
Bod varu: 224…226 °C
Bod tání: 2 °C
Hustota: 1,322
Bod vzplanutí: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000073
CAS číslo: 576-83-0
EINECS: 209-405-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR