1,3-Diethylbenzene ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Purity: 97+%
CAS No.: 141-93-5
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 55°(131°F)
Physical Form: Liquid
Density: 0.86
Refractive Index: 1.4960
Appearance: Colourless
Odour: Aromatic
Boiling Point: 182°
UN No.: 2049
MDL No.: MFCD00009260
Vzorec:
C₁₀H₁₄ Bod varu: 182 °C Hustota: 0,86 Bod vzplanutí: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009260 CAS číslo: 141-93-5 EINECS: 205-511-4 UN: 2049 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...