2,6-Dimethylthiophenol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1,3-Dimethyl-2-mercaptobenzene, 2-Mercapto-m-xylene, 2,6-Dimethylbenzene-1-thiol, 2,6-Dimethylbenzenethiol
A13286.03
A13286.06
A13286.14
A13286.03EA
1290
CZK
A13286.03
A13286.06
A13286.14
2,6-Dimethylthiophenol ≥97%
2,6-Dimethylthiophenol
Purity: 97%
CAS No.: 118-72-9
Molecular Formula: C8H10S
Formula Weight: 138.23
Flash Point: 85°(185°F)
Refractive Index: 1.5749
Odour: Unpleasant
Boiling Point: 122°/50mm
Beilstein Reference: 1099405
UN No.: 2810
MDL No.: MFCD00010021
Vzorec:
C₈H₁₀S MW: 138.23 Bod varu: 122 °C (50 mmHg) Hustota: 1.038 Bod vzplanutí: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010021 CAS číslo: 118-72-9 EINECS: 204-272-3 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...