Tisk…

2,6-Xylenol ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene

A11947.0E A11947.36
A11947.0EEA 5120 CZK
A11947.0E A11947.36
2,6-Xylenol ≥99%
2,6-Xylenol

Purity: 99% CAS No.: 576-26-1 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 78°(172°F) Melting Point: 44-48° Boiling Point: 203° Beilstein Reference: 1446677 Merck Reference: 14,10082 UN No.: 2261 MDL No.: MFCD00002240

Vzorec: C₈H₁₀O
MW: 122.17
Bod varu: 203 °C
Bod tání: 44…48 °C
Hustota: 1.15
Bod vzplanutí: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002240
CAS číslo: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1 II
Merck Index: 12,10215

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR