Tisk…

2,6-Di-tert-butylphenol ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1,3-Di(dimethylethyl)-2-hydroxybenzene

A17244.0B A17244.0I A17244.30
A17244.0BEA 2080 CZK
A17244.0B A17244.0I A17244.30
2,6-Di-tert-butylphenol ≥98%
2,6-Di-tert-butylphenol

Purity: 98% CAS No.: 128-39-2 Molecular Formula: C14H22O Formula Weight: 206.33 Flash Point: 120°(248°F) Odour: Phenol-like Boiling Point: 252-253° Beilstein Reference: 1841887 UN No.: 3077 MDL No.: MFCD00008820

Vzorec: C₁₄H₂₂O
MW: 206,33 g/mol
Bod varu: 252…253 °C
Bod tání: 35…38 °C
Hustota: 0,914
Bod vzplanutí: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008820
CAS číslo: 128-39-2
EINECS: 204-884-0
UN: 3077
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR