Tisk…

4-Nitro-o-xylene ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3,4-Dimethyl-1-nitrobenzene, 3,4-Dimethylnitrobenzene, 1,2-Dimethyl-4-nitrobenzene, 4-Nitro-o-xylene

L05342.22 L05342.36
L05342.22EA 1150 CZK
L05342.22 L05342.36
4-Nitro-o-xylene ≥99%
4-Nitro-o-xylene

Purity: 99% CAS No.: 99-51-4 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 113°(235°F) Refractive Index: 1.5560 Melting Point: 27-31° Boiling Point: 255-257° Beilstein Reference: 2043245 UN No.: 3447 MDL No.: MFCD00007268

Vzorec: C₈H₉NO₂
MW: 151.16
Bod varu: 255…257 °C
Bod tání: 27…31 °C
Hustota: 1.136
Bod vzplanutí: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007268
CAS číslo: 99-51-4
EINECS: 202-761-6
UN: 3447
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR