2,6-Xylidine ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Amino-m-xylene, 2,6-Dimethylaniline
A14512.0B
A14512.22
A14512.36
A14512.0BEA
3780
CZK
A14512.0B
A14512.22
A14512.36
2,6-Xylidine ≥99%
2,6-Xylidine
Purity: 99%
CAS No.: 87-62-7
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 91°(195°F)
Refractive Index: 1.5600
Odour: Characteristic
Melting Point: 11°
Boiling Point: 216°
Beilstein Reference: 636332
Merck Reference: 14,10084
UN No.: 1711
MDL No.: MFCD00007747
Vzorec:
C₈H₁₁N Bod varu: 216 °C Bod tání: 11 °C Hustota: 0,981 Bod vzplanutí: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007747 CAS číslo: 87-62-7 EINECS: 201-758-7 UN: 1711 ADR: 6.1,II Merck Index: 14,10084 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...