2,5-Xylidine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-p-xylene, 2,5-Dimethylaniline
A12682.0E
A12682.22
A12682.36
A12682.14
A12682.0EEA
0
CZK
A12682.0E
A12682.22
A12682.36
A12682.14
2,5-Xylidine ≥98%
2,5-Xylidine
Purity: 98+%
CAS No.: 95-78-3
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 93°(199°F)
Refractive Index: 1.5590
Odour: Characteristic
Melting Point: 15°
Boiling Point: 218°
Beilstein Reference: 2205178
Merck Reference: 14,10084
UN No.: 1711
MDL No.: MFCD00007743
Vzorec:
C₈H₁₁N MW: 121.18 Bod varu: 218 °C Bod tání: 15 Hustota: 0.973 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007743 CAS číslo: 95-78-3 EINECS: 202-451-0 UN: 1711 ADR: 6.1 II Merck Index: 12,10217 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...