4-tert-Butylbenzylamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
p-tert-Butylbenzylamine
L20394.06
L20394.14
L20394.06EA
2220
CZK
L20394.06
L20394.14
4-tert-Butylbenzylamine 98%
4-tert-Butylbenzylamine
Purity: 98%
CAS No.: 39895-55-1
Molecular Formula: C11H17N
Formula Weight: 163.26
Flash Point: 107°(224°F)
Refractive Index: 1.5210
Boiling Point: 124°/16mm
Beilstein Reference: 2961797
UN No.: 2735
MDL No.: MFCD00040754
Vzorec:
C₁₁H₁₇N MW: 163,26 g/mol Bod varu: 124 °C (16 mmHg) Hustota: 0,927 Bod vzplanutí: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040754 CAS číslo: 39895-55-1 EINECS: 254-681-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...