4-Iodo-m-xylene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2,4-Dimethyliodobenzene, 1-Iodo-2,4-dimethylbenzene, 2,4-Dimethyl-1-iodobenzene, 1,3-Dimethyl-4-iodobenzene
A16878.09
A16878.18
A16878.09EA
1200
CZK
A16878.09
A16878.18
4-Iodo-m-xylene ≥98%
4-Iodo-m-xylene
Purity: 98%
CAS No.: 4214-28-2
Molecular Formula: C8H9I
Formula Weight: 232.07
Flash Point:
Vzorec:
C₈H₉I Bod varu: 231…232 °C Hustota: 1,628 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013706 CAS číslo: 4214-28-2 EINECS: 224-141-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...