α-Chloro-m-xylene ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride
A10784.14
A10784.22
A10784.36
A10784.14EA
1030
CZK
A10784.14
A10784.22
A10784.36
α-Chloro-m-xylene ≥98%
α-Chloro-m-xylene
Purity: 98%
CAS No.: 620-19-9
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Flash Point: 75°(167°F)
Refractive Index: 1.5350
Odour: Pungent
Boiling Point: 195-196°
Beilstein Reference: 2040357
Merck Reference: 14,10091
UN No.: 3265
MDL No.: MFCD00000909
Vzorec:
C₈H₉Cl MW: 140,61 g/mol Bod varu: 195…196 °C Hustota: 1,066 Bod vzplanutí: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000909 CAS číslo: 620-19-9 EINECS: 210-628-9 UN: 3265 ADR: 8,II Merck Index: 14,10091 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...