2,5-Dichloro-p-xylene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B21344.14
B21344.22
B21344.14EA
2000
CZK
B21344.14
B21344.22
2,5-Dichloro-p-xylene ≥98%
2,5-Dichloro-p-xylene
Purity: 98%
CAS No.: 1124-05-6
Molecular Formula: C8H8Cl2
Formula Weight: 175.06
Melting Point: 68-70°
Boiling Point: 221-223°
MDL No.: MFCD00000610
Vzorec:
C₈H₈Cl₂ Bod varu: 221…223 °C Bod tání: 68…70 °C Hustota: 1,39 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000610 CAS číslo: 1124-05-6 EINECS: 214-387-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...