Tisk…

(+)-Valencene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W344303-100G-K W344303-1KG-K W344303-25G-K
SIALW344303-100G-KEA 4740 CZK
SIALW344303-100G-K SIALW344303-1KG-K SIALW344303-25G-K
(+)-Valencene, Sigma-Aldrich®
(+)-Valencene
Vzorec: C₁₅H₂₄
MDL Number: MFCD00075884
CAS číslo: 4630-07-3
EINECS: 225-047-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR