Tisk…

Benzo[b]thiophene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Thianaphthene, 1-Benzothiophene

M-Clarity™ quality level = MQ100

T27405-100G T27405-25G
SIALT27405-100GEA 12240 CZK
SIALT27405-100G SIALT27405-25G
Benzo[b]thiophene, Sigma-Aldrich®
Benzo[b]thiophene
Vzorec: C₈H₆S
MW: 134,2 g/mol
MDL Number: MFCD00005864
CAS číslo: 95-15-8
EINECS: 202-395-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR