Tisk…

1-Phenyl-2-urea, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Phenylurea, 1-Phenylurea, N-Phenylurea

M-Clarity™ quality level = MQ200

P36959-100G P36959-500G
SIALP36959-100GEA 1110 CZK
SIALP36959-100G SIALP36959-500G
1-Phenyl-2-urea, Sigma-Aldrich®
1-Phenyl-2-urea
Vzorec: C₇H₈N₂O
MW: 136,15 g/mol
MDL Number: MFCD00007944
CAS číslo: 64-10-8
EINECS: 200-576-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR