Farnesol (mixture of isomers), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
F203-5G
F203-100G
F203-25G
SIALF203-5GEA
649
CZK
SIALF203-5G
SIALF203-100G
SIALF203-25G
Farnesol (mixture of isomers), Sigma-Aldrich®
Farnesol (mixture of isomers)
Vzorec:
C₁₅H₂₆O |
MDL Number:
MFCD00002918 CAS číslo: 4602-84-0 EINECS: 225-004-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...