2-Chloroethylammonium chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
C40200-100G
C40200-5G
SIALC40200-100GEA
1390
CZK
SIALC40200-100G
SIALC40200-5G
2-Chloroethylammonium chloride, Sigma-Aldrich®
2-Chloroethylammonium chloride
Vzorec:
C₂H₇Cl₂N |
MDL Number:
MFCD00012887 CAS číslo: 870-24-6 EINECS: 212-793-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...