Tisk…

Benzoin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Hydroxy-2-phenylacetophenone

M-Clarity™ quality level = MQ100

B8681-100G B8681-500G
SIALB8681-100GEA 1110 CZK
SIALB8681-100G SIALB8681-500G
Benzoin, Sigma-Aldrich®
Benzoin
Vzorec: C₁₄H₁₂O₂
MW: 212,25 g/mol
MDL Number: MFCD00004496
CAS číslo: 119-53-9
EINECS: 204-331-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR