Tisk…

Benzyldimethyldodecylammonium bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Benzyldodecyldimethylammonium bromide, Benzalkonium bromide, N-Benzyl-N,N-dimethyldodecan-1-aminium bromide

M-Clarity™ quality level = MQ200

B5776-10G B5776-50G
SIALB5776-10GEA 3860 CZK
SIALB5776-10G SIALB5776-50G
Benzyldimethyldodecylammonium bromide, Sigma-Aldrich®
Benzyldimethyldodecylammonium bromide
Vzorec: C₂₁H₃₈BrN
MW: 384,44 g/mol
MDL Number: MFCD00011768
CAS číslo: 7281-04-1
EINECS: 230-698-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR