Benzylammonium chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Benzylamine hydrochloride
M-Clarity™ quality level = MQ200
B5136-100G
B5136-25G
SIALB5136-100GEA
2510
CZK
SIALB5136-100G
SIALB5136-25G
Benzylammonium chloride, Sigma-Aldrich®
Benzylammonium chloride
Vzorec:
C₇H₁₀ClN |
MDL Number:
MFCD00012852 CAS číslo: 3287-99-8 EINECS: 221-943-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...