Tisk…

4-Aminomorpholine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

A66308-10G A66308-25G
SIALA66308-10GEA 3330 CZK
SIALA66308-10G SIALA66308-25G
4-Aminomorpholine, Sigma-Aldrich®
4-Aminomorpholine
Vzorec: C₄H₁₀N₂O
MDL Number: MFCD00006174
CAS číslo: 4319-49-7
EINECS: 224-347-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR