Tisk…

N-(2-Aminoethyl)morpholine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(2-Aminoethyl)morpholine, 2-Morpholinoethanamine

M-Clarity™ quality level = MQ200

A55004-100G A55004-25G A55004-5G
SIALA55004-100GEA 6750 CZK
SIALA55004-100G SIALA55004-25G SIALA55004-5G
N-(2-Aminoethyl)morpholine, Sigma-Aldrich®
N-(2-Aminoethyl)morpholine
Vzorec: C₆H₁₄N₂O
MW: 130,19 g/mol
MDL Number: MFCD00006182
CAS číslo: 2038-03-1
EINECS: 218-011-6
UN: 2923
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR