Tisk…
Perfluorodecahydronaphthalene (mixture of cis and trans isomers), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Perfluoro(decahydronaphthalene), Perfluorodecalin, Perfluorodecahydronaphthalene
M-Clarity™ quality level = MQ100
77264-10ML 77264-250ML 77264-50ML
SIAL77264-10MLEA 2090 CZK
SIAL77264-10ML SIAL77264-250ML SIAL77264-50ML
Perfluorodecahydronaphthalene (mixture of cis and trans isomers), Sigma-Aldrich®
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Vzorec: C₁₀F₁₈
MW: 462,08 g/mol
MDL Number: MFCD00010626
CAS číslo: 306-94-5
EINECS: 206-192-4