Tisk…

(R)-(+)-4-Fluoro-α-methylbenzylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (R)-(+)-1-(4-Fluorophenyl)ethylamine, (R)-1-(4-Fluorophenyl)ethanamine

727024-25G 727024-5G
SIAL727024-25GEA 0 CZK
SIAL727024-25G SIAL727024-5G
(R)-(+)-4-Fluoro-α-methylbenzylamine, Sigma-Aldrich®
(R)-(+)-4-Fluoro-α-methylbenzylamine
Vzorec: C₈H₁₀FN
MW: 139,17 g/mol
MDL Number: MFCD03093090
CAS číslo: 374898-01-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR