Tisk…

3-Aminobenzylamin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: alpha-Amino-m-toluidine, 3-(Aminomethyl)aniline

721492-25G 721492-5G
SIAL721492-25GEA 0 CZK
SIAL721492-25G SIAL721492-5G
3-Aminobenzylamin, Sigma-Aldrich®
3-Aminobenzylamin
Vzorec: C₇H₁₀N₂
MW: 122,17 g/mol
MDL Number: MFCD00078355
CAS číslo: 4403-70-7
EINECS: 412-230-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR