Tisk…

3-Nitrophenethyl bromide, Sigma-Aldrich®

Dodavatel: Merck

691984-1G 691984-5G
SIAL691984-1GEA 1820 CZK
SIAL691984-1G SIAL691984-5G
3-Nitrophenethyl bromide, Sigma-Aldrich®
3-Nitrophenethyl bromide
Vzorec: C₈H₈BrNO₂
MW: 230,06 g/mol
CAS číslo: 16799-04-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR