Tisk…

4-Methoxyphenethyl bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Bromo-2-(4-methoxyphenyl)ethane, 1-(2-Bromoethyl)-4-methoxybenzene, 4-(2-Bromoethyl)anisole

M-Clarity™ quality level = MQ100

655155-1G 655155-5G
SIAL655155-1GEA 1580 CZK
SIAL655155-1G SIAL655155-5G
4-Methoxyphenethyl bromide, Sigma-Aldrich®
4-Methoxyphenethyl bromide
Vzorec: C₉H₁₁BrO
MW: 215,09 g/mol
MDL Number: MFCD07369786
CAS číslo: 14425-64-0
EINECS: 238-393-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR