Tisk…

4-Bromophenethyl bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Bromo-4-(2-bromoethyl)benzene

M-Clarity™ quality level = MQ100

651982-10G 651982-1G
SIAL651982-10GEA 0 CZK
SIAL651982-10G SIAL651982-1G
4-Bromophenethyl bromide, Sigma-Aldrich®
4-Bromophenethyl bromide
Vzorec: C₈H₈Br₂
MW: 263,96 g/mol
MDL Number: MFCD00045052
CAS číslo: 1746-28-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR