Tisk…

4-Methyl-2-thenoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Methyl-2-thiophenecarboxylic acid, 4-Methylthiophene-2-carboxylic acid

M-Clarity™ quality level = MQ100

633550-25G 633550-5G
SIAL633550-25GEA 10570 CZK
SIAL633550-25G SIAL633550-5G
4-Methyl-2-thenoic acid, Sigma-Aldrich®
4-Methyl-2-thenoic acid
Vzorec: C₆H₆O₂S
MW: 142,18 g/mol
MDL Number: MFCD00089284
CAS číslo: 14282-78-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR