Tisk…

Dichloro(diphenyl)silane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diphenyldichlorosilane, Dichlorodiphenylsilane

M-Clarity™ quality level = MQ100

440124-100ML 440124-500ML
SIAL440124-100MLEA 1840 CZK
SIAL440124-100ML SIAL440124-500ML
Dichloro(diphenyl)silane, Sigma-Aldrich®
Dichloro(diphenyl)silane
Vzorec: C₁₂H₁₀Cl₂Si
MW: 253.2
MDL Number: MFCD00000489
CAS číslo: 80-10-4
EINECS: 201-251-0
UN: 1769
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR