2-Aminobenzylamin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(Aminomethyl)aniline
M-Clarity™ quality level = MQ100
348015-10G
348015-50G
SIAL348015-10GEA
3460
CZK
SIAL348015-10G
SIAL348015-50G
2-Aminobenzylamin, Sigma-Aldrich®
2-Aminobenzylamin
Vzorec:
C₇H₁₀N₂ MW: 122,17 g/mol |
MDL Number:
MFCD00075408 CAS číslo: 4403-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...