(1S,2R)-(+)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
331880-1G
331880-5G
SIAL331880-1GEA
1860
CZK
SIAL331880-1G
SIAL331880-5G
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
Vzorec:
C₁₄H₁₅NO MW: 213,28 g/mol |
MDL Number:
MFCD00074959 CAS číslo: 23364-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...