Tisk…

trans,trans-Farnesol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-ol

M-Clarity™ quality level = MQ200

277541-10G 277541-1G
SIAL277541-10GEA 5530 CZK
SIAL277541-10G SIAL277541-1G
trans,trans-Farnesol, Sigma-Aldrich®
trans,trans-Farnesol
Vzorec: C₁₅H₂₆O
MDL Number: MFCD00002918
CAS číslo: 106-28-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR