Tisk…

Triethylaluminium 1 mol/l in hexane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Triethylaluminum

M-Clarity™ quality level = MQ100

252662-100ML 252662-800ML
SIAL252662-100MLEA 3310 CZK
SIAL252662-100ML SIAL252662-800ML
Triethylaluminium 1 mol/l in hexane, Sigma-Aldrich®
Triethylaluminium
Vzorec: C₆H₁₅Al
MW: 114.17
Bod varu: 128
Bod tání: -46.1
MDL Number: MFCD00009015
CAS číslo: 97-93-8
EINECS: 202-619-3
UN: 3394
ADR: 4.2 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR