Tisk…

4-Formylmorpholine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Morpholine-4-carbaldehyde, N-Formylmorpholine

MQ200

250376-100G 250376-500G
SIAL250376-100GEA 1380 CZK
SIAL250376-100G SIAL250376-500G
4-Formylmorpholine, Sigma-Aldrich®
4-Formylmorpholine
Vzorec: C₅H₉NO₂
MW: 115.13
MDL Number: MFCD00006170
CAS číslo: 4394-85-8
EINECS: 224-518-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR