Tisk…

Sodium bis(trimethylsilyl)amide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: HMDS sodium salt, N-Sodiohexamethyldisilazane, Sodium bis(trimethylsilyl)amide, HMDS-Na

M-Clarity™ quality level = MQ100

235083-100G 235083-25G 235083-5G
SIAL235083-100GEA 11930 CZK
SIAL235083-100G SIAL235083-25G SIAL235083-5G
Sodium bis(trimethylsilyl)amide, Sigma-Aldrich®
Sodium bis(trimethylsilyl)amide
Vzorec: C₆H₁₈NNaSi₂
MW: 183,38 g/mol
Hustota: 1,065 g/cm³ (20 °C)
MDL Number: MFCD00009835
CAS číslo: 1070-89-9
EINECS: 213-983-8
UN: 3096
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR