Tisk…

Selenourea, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

230499-10G 230499-1G
SIAL230499-10GEA 19140 CZK
SIAL230499-10G SIAL230499-1G
Selenourea, Sigma-Aldrich®
Selenourea
Vzorec: CH₄N₂Se
MDL Number: MFCD00008065
CAS číslo: 630-10-4
EINECS: 211-129-9
UN: 3283
ADR: 6.1,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR