Tisk…
N-Methylmorpholine-N-oxide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide
M-Clarity™ quality level = MQ200
224286-25G 224286-100G 224286-5G
SIAL224286-25GEA 2380 CZK
SIAL224286-25G SIAL224286-100G SIAL224286-5G
N-Methylmorpholine-N-oxide, Sigma-Aldrich®
N-Methylmorpholine-N-oxide
Vzorec: C₅H₁₁NO₂
MW: 117.15
MDL Number: MFCD00005947
CAS číslo: 7529-22-8
EINECS: 231-391-8
UN: 3082
ADR: 9 III