Tisk…

4-Hexyloxyaniline, Sigma-Aldrich®

Dodavatel: Merck


MQ100

222100-25G 222100-5G
SIAL222100-25GEA 9580 CZK
SIAL222100-25G SIAL222100-5G
4-Hexyloxyaniline, Sigma-Aldrich®
4-Hexyloxyaniline
Vzorec: C₁₂H₁₉NO
MW: 193.29
MDL Number: MFCD00007868
CAS číslo: 39905-57-2
EINECS: 254-696-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR