Tisk…

tert-Butylchlorodiphenylsilane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane

M-Clarity™ quality level = MQ200

195537-10G 195537-50G 195537-2G
SIAL195537-10GEA 1580 CZK
SIAL195537-10G SIAL195537-50G SIAL195537-2G
tert-Butylchlorodiphenylsilane, Sigma-Aldrich®
tert-Butylchlorodiphenylsilane
Vzorec: C₁₆H₁₉ClSi
MDL Number: MFCD00000497
CAS číslo: 58479-61-1
EINECS: 261-282-0
UN: 2987
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR