2-Methoxypropene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Methoxy-1-propene, Isopropenyl methyl ether, 2-Methoxyprop-1-ene
M-Clarity™ quality level = MQ100
174645-250G
174645-50G
SIAL174645-250GEA
3400
CZK
SIAL174645-250G
SIAL174645-50G
2-Methoxypropene, Sigma-Aldrich®
2-Methoxypropene
Vzorec:
C₄H₈O |
MDL Number:
MFCD00014929 CAS číslo: 116-11-0 EINECS: 204-125-3 UN: 1993 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...