Tisk…

(±)-4-Fluoro-α-methylbenzylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: DL-4-Fluoro-α-methylbenzylamine, 1-(4-Fluorophenyl)ethylamine

M-Clarity™ quality level = MQ200

154040-1G 154040-5G
SIAL154040-1GEA 1400 CZK
SIAL154040-1G SIAL154040-5G
(±)-4-Fluoro-α-methylbenzylamine, Sigma-Aldrich®
(±)-4-Fluoro-α-methylbenzylamine
Vzorec: C₈H₁₀FN
MW: 139,17 g/mol
MDL Number: MFCD00041323
CAS číslo: 403-40-7
EINECS: 206-958-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR