Tisk…

4-Methylphenethylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(p-Tolyl)ethylamine

M-Clarity™ quality level = MQ100

132020-1G 132020-5G
SIAL132020-1GEA 1620 CZK
SIAL132020-1G SIAL132020-5G
4-Methylphenethylamine, Sigma-Aldrich®
4-Methylphenethylamine
Vzorec: C₉H₁₃N
MW: 135,21 g/mol
MDL Number: MFCD00008195
CAS číslo: 3261-62-9
EINECS: 221-865-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR