Tisk…

Hydrogen peroxide-urea, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Urea hydrogen peroxide

M-Clarity™ quality level = MQ200

04078-100G 04078-500G
SIAL04078-100GEA 1880 CZK
SIAL04078-100G SIAL04078-500G
Hydrogen peroxide-urea, Sigma-Aldrich®
Hydrogen peroxide-urea
Vzorec: CH₆N₂O₃
MW: 94,07 g/mol
MDL Number: MFCD00013119
CAS číslo: 124-43-6
EINECS: 204-701-4
UN: 1511
ADR: 5.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR