2-Chloroethylammonium chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
8.20265.0100
8.20265.0250
8.20265.0100EA
1070
CZK
8.20265.0100
8.20265.0250
2-Chloroethylammonium chloride, Sigma-Aldrich®
2-Chloroethylammonium chloride
Vzorec:
C₂H₇Cl₂N Storage Temperature: Ambient |
MDL Number:
MFCD00012887 CAS číslo: 870-24-6 EINECS: 212-793-2 UN: 3261 ADR: 8,III |
Výsledky specifikačního testu
Solubility | 5666 g/l (20 °C) |
Melting Point | 143 - 146 °C |
Molar Mass | 115.99 g/mol |
Bulk Density | 750 - 850 kg/m3 |
pH | 2 - 3 (5666 g/l, H2O, 20 °C) |
Synonyms | 2-Chloroethylamine hydrochloride, ß-Aminoethyl chloride hydrochloride |
Hill Formula | C2H7Cl2N |
Chemical Formula | ClCH2CH2NH2 * HCl |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...