Tisk…
Lidocaine, powder_white
Dodavatel: MP Biomedicals
Varování
0219011125 0219011180
ICNA0219011125EA 1780 CZK
ICNA0219011125 ICNA0219011180
Lidocaine, powder_white
Lidocaine
It is Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.
Lidocaine is used to treat allergies. It is generally used to differentiate and identify leucocytes, malaria parasites, and trypanosomas.
It is Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.
Soluble to 100 mM in ethanol.
Upozornení: Research Use Only (RUO).
Vzorec: C₁₄H₂₂N₂O
MW: 234.34
Bod tání: 68.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00026733
CAS číslo: 137-58-6
EINECS: 205-302-8
Objednat
Výsledky specifikačního testu
Identity Test Conforms to Standard