2,4-Dimethylbenzoic acid 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A10589.09
A10589.18
A10589.09EA
1330
CZK
A10589.09
A10589.18
2,4-Dimethylbenzoic acid 97%
2,4-Dimethylbenzoic acid
Purity: 98%
CAS No.: 611-01-8
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 154°(309°F)
Odour: Odourless
Boiling Point: 267°/727mm
Beilstein Reference: 971589
MDL No.: MFCD00002480
Vzorec:
C₉H₁₀O₂ Bod varu: 267 °C (727 mmHg) Bod tání: 124…126 °C Bod vzplanutí: 154 °C (309 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002480 CAS číslo: 611-01-8 EINECS: 210-246-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...