Tisk…

2,3-Dimethylbenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

A10383.06 A10383.14 A10383.22
A10383.06EA 1610 CZK
A10383.06 A10383.14 A10383.22
2,3-Dimethylbenzoic acid ≥98%
2,3-Dimethylbenzoic acid

Purity: 98% CAS No.: 603-79-2 Molecular Formula: C9H10O2 Formula Weight: 150.18 Odour: Odourless Melting Point: 143-147° Beilstein Reference: 971590 MDL No.: MFCD00002479

Vzorec: C₉H₁₀O₂
MW: 150,18 g/mol
Bod varu: 282 °C (1013 hPa)
Bod tání: 143…147 °C
Hustota: 1,283 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002479
CAS číslo: 603-79-2
EINECS: 210-058-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR