3-Formyl-2-thiopheneboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Formylthiophene-2-boronic acid, (3-Formylthiophen-2-yl)boronic acid
L15196.03
L15196.06
L15196.03EA
5430
CZK
L15196.03
L15196.06
3-Formyl-2-thiopheneboronic acid ≥97%
3-Formyl-2-thiopheneboronic acid
Purity: 97%
CAS No.: 17303-83-2
Molecular Formula: C5H5BO3S
Formula Weight: 155.97
Storage Temperature: 2°C to 8°C
Melting Point: ca 180° dec.
Beilstein Reference: 4387740
MDL No.: MFCD01075679
Vzorec:
C₅H₅BO₃S MW: 155,97 g/mol Bod tání: ∼ 180 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD01075679 CAS číslo: 17303-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...